Assay Details
| Gene Symbol: 4832441B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014650.1 | - | 1 | 854 | 81 | |
| AK029342.1 | - | 1 | 854 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 24 (sodium/potassium/calcium exchanger), member 2 |
Slc24a2 |
2810021B17Rik, 6330417K15Rik, AI847460, Nckx2 |
-566.5kb | Sense | Chr. 4 - 86983124 - 87230554 on NCBI Build GRCm38 |
| Downstream | focadhesin | Focad |
Kiaa1797, mKIAA1797 |
295.9kb | Anti-sense | Chr. 4 - 88094630 - 88411011 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4832441B07Rik
Genomic Map