Assay Details
| Gene Symbol: 4833412K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014687.1 | - | 1 | 948 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phenylalanyl-tRNA synthetase, beta subunit |
Farsb |
C76708, Farsa, Farsl, Farslb, Frsb |
Sense | Chr. 1 - 78417958 - 78488897 on NCBI Build GRCm38 | |
| Downstream | monoacylglycerol O-acyltransferase 1 | Mogat1 |
0610030A14Rik, 1110064N14Rik, Dgat2l, Dgat2l1, MGAT1, WI1-2612I11.1, mDC2 |
44.1kb | Anti-sense | Chr. 1 - 78511060 - 78538173 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4833412K13Rik
Genomic Map