Assay Details
| Gene Symbol: 4833417J20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014716.1 | - | 1 | 1154 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
carnitine palmitoyltransferase 1a, liver |
Cpt1a |
C730027G07, CPTI, Cpt1 |
-476.0 | Sense | Chr. 19 - 3322326 - 3385735 on NCBI Build GRCm38 |
| Downstream |
metallothionein-like 5, testis-specific (tesmin) |
Mtl5 | tesmin | 861.0 | Sense | Chr. 19 - 3388857 - 3407823 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4833417J20Rik
Genomic Map