Assay Details
Gene Symbol: 4930403O18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015070.1 | - | 1 | 1054 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930570E03 gene | 4930570E03Rik | Sense | Chr. 16 - 78694989 - 78761063 on NCBI Build GRCm38 | ||
Downstream | chondrolectin | Chodl |
3110074E07Rik, MT75, PRED12 |
171.9kb | Sense | Chr. 16 - 78930864 - 78951733 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
