Assay Details
Gene Symbol: Tcte2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U46151.1 | - | 2 - 3 | 411 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 16046 | Gm16046 | OTTMUSG00000029502 | -37.0kb | Sense | Chr. 17 - 13679250 - 13685440 on NCBI Build GRCm38 |
Downstream |
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4 |
Mllt4 |
5033403D15Rik, AF6, Af-6, Afadin, Gm314, I-afadin, S-afadin |
32.5kb | Anti-sense | Chr. 17 - 13760541 - 13905798 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
