Assay Details
Gene Symbol: Rara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | X56570.1 | - | 1 - 2 | 73 | 53 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cell division cycle 6 | Cdc6 | CDC18L | -40.1kb | Sense | Chr. 11 - 98907702 - 98923942 on NCBI Build GRCm38 |
Downstream |
gap junction protein, delta 3 |
Gjd3 |
Gja11, Gjc1, cx30.2 |
15.8kb | Anti-sense | Chr. 11 - 98982180 - 98983016 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
