Assay Details
Gene Symbol: 2610507B11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | X81717.1 | - | 1 - 2 | 114 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | stromal cell derived factor 2 | Sdf2 | AI853825 | -23.2kb | Anti-sense | Chr. 11 - 78245746 - 78255496 on NCBI Build GRCm38 |
Downstream | cDNA sequence BC030499 | BC030499 |
BC037104, Sgk494 |
11.7kb | Anti-sense | Chr. 11 - 78290732 - 78296805 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
