Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200008146.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008146.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008146.mm.1 |
TC1200001142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008155.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008155.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008155.mm.1 |
TC1200001142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008148.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008148.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008148.mm.1 |
TC1200001142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008145.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008145.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008145.mm.1 |
TC1200001142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008152.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008152.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008152.mm.1 |
TC1200001142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008154.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008154.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008154.mm.1 |
TC1200001142.mm.1 |
MTA TC |