Assay Details
| Gene Symbol: Tmed4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003613.1 | - | 1 | 847 | 133 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 56 | Ddx56 |
2600001H07Rik, D11Ertd619e, Noh61 |
-2.6kb | Sense | Chr. 11 - 6257545 - 6267729 on NCBI Build GRCm38 |
| Downstream | oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide) | Ogdh |
2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192 |
20.1kb | Anti-sense | Chr. 11 - 6291593 - 6359094 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmed4
Genomic Map