Assay Details
| Gene Symbol: Synpo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004418.1 | - | 1 | 944 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myozenin 2 | Myoz2 |
1110012I24Rik, Fatz-2, Myozl2 |
-76.0kb | Sense | Chr. 3 - 123006206 - 123035001 on NCBI Build GRCm38 |
| Downstream |
Sec24 related gene family, member D (S. cerevisiae) |
Sec24d |
2310020L09Rik, Gm1349 |
155.4kb | Anti-sense | Chr. 3 - 123267496 - 123365641 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Synpo2
Genomic Map