Assay Details
| Gene Symbol: 1700123O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045799.1 | 1 - 2 | 254 | 83 | ||
| GenBank mRNA | AK006584.1 | - | 1 - 2 | 255 | 83 | |
| AK018984.1 | - | 1 - 2 | 256 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36902 |
Gm36902 | -24.4kb | Anti-sense | Chr. 16 - 5946938 - 5951145 on NCBI Build GRCm38 | |
| Downstream | transmembrane protein 114 | Tmem114 |
4930511J11Rik, Cldn26 |
2.4mb | Anti-sense | Chr. 16 - 8409276 - 8425136 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700123O21Rik
Genomic Map