Assay Details
Gene Symbol: 1700092C02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045467.1 | 2 - 3 | 226 | 84 | ||
GenBank mRNA | AK007037.1 | - | 2 - 3 | 227 | 84 | |
AK018991.1 | - | 2 - 3 | 227 | 84 | ||
BY707399.1 | - | 2 - 3 | 227 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 184C | Tmem184c |
8430433H16Rik, AI645453, AL033298, BC004056, Tmem34 |
-16.8kb | Anti-sense | Chr. 8 - 77595978 - 77610759 on NCBI Build GRCm38 |
Downstream | endothelin receptor type A | Ednra |
ET-AR, ETa, Gpcr10 |
34.1kb | Anti-sense | Chr. 8 - 77663029 - 77724452 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
