Assay Details
| Gene Symbol: 4921501M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019533.1 | - | 1 | 652 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC100451 | BC100451 |
Ddc8, FS39, Kiaa1731nl |
-39.0 | Anti-sense | Chr. 11 - 118332360 - 118342568 on NCBI Build GRCm38 |
| Downstream |
lectin, galactoside-binding, soluble, 3 binding protein |
Lgals3bp |
90K, CyCAP, MAC-2BP, Ppicap, Tango10b |
49.2kb | Anti-sense | Chr. 11 - 118392752 - 118401931 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4921501M06Rik
Genomic Map