Assay Details
Gene Symbol: 4930448L02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019618.1 | - | 1 | 148 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
proteasome (prosome, macropain) activator subunit 4 |
Psme4 |
AA409398, AU041366, TEMO, mKIAA0077 |
Anti-sense | Chr. 11 - 30771328 - 30881150 on NCBI Build GRCm38 | |
Downstream | endoplasmic reticulum lectin 1 | Erlec1 | 4933407N01Rik | 94.0kb | Sense | Chr. 11 - 30865632 - 30954173 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
