Assay Details
| Gene Symbol: 1810019N24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019651.1 | - | 1 | 51 | 85 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY715158.1 | - | 1 | Chr. 7 - 12949690 - 12950066 [-] on Build GRCm38 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 225 | Rnf225 |
2310014L17Rik, AV237412, C77118 |
-18.6kb | Anti-sense | Chr. 7 - 12927416 - 12931072 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 324 | Zfp324 |
A930002M05, D430030K24Rik, ZF5128 |
15.7kb | Anti-sense | Chr. 7 - 12965811 - 12974244 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1810019N24Rik
Genomic Map