Assay Details
Gene Symbol: 4930486F22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038024.1 | 1 - 2 | 29 | 84 | ||
GenBank mRNA | AK019657.1 | - | 1 - 2 | 25 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | synapsin III | Syn3 | Anti-sense | Chr. 10 - 86048746 - 86499019 on NCBI Build GRCm38 | ||
Downstream | tissue inhibitor of metalloproteinase 3 | Timp3 | Timp-3 | 193.1kb | Sense | Chr. 10 - 86300412 - 86349505 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
