Assay Details
Gene Symbol: 4930528J18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019708.1 | - | 1 | 349 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930423D24 gene | 4930423D24Rik | -8.7kb | Anti-sense | Chr. 10 - 117177625 - 117181110 on NCBI Build GRCm38 | |
Downstream | YEATS domain containing 4 | Yeats4 |
4930573H17Rik, B230215M10Rik, GAS41, NuBI-1 |
24.9kb | Sense | Chr. 10 - 117215141 - 117224507 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
