Assay Details
| Gene Symbol: 4930549L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019755.1 | - | 1 | 347 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 7 (cationic amino acid transporter, y+ system), member 1 |
Slc7a1 |
4831426K01Rik, AI447493, Atrc-1, Atrc1, Cat1, Rec-1, Rev-1, mCAT-1 |
-90.0kb | Anti-sense | Chr. 5 - 148327410 - 148399904 on NCBI Build GRCm38 |
| Downstream | ubiquitin-like 3 | Ubl3 |
AW108023, HCG |
14.2kb | Anti-sense | Chr. 5 - 148504631 - 148552788 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930549L11Rik
Genomic Map