Assay Details
| Gene Symbol: 6030451C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020069.1 | - | 1 | 353 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein kinase, AMP-activated, alpha 2 catalytic subunit |
Prkaa2 |
2310008I11Rik, A830082D05, AMPKalpha2 |
-90.8kb | Anti-sense | Chr. 4 - 105029650 - 105109898 on NCBI Build GRCm38 |
| Downstream | predicted gene 12724 | Gm12724 | OTTMUSG00000008145 | 1.0mb | Sense | Chr. 4 - 106237122 - 106291000 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6030451C04Rik
Genomic Map