Assay Details
Gene Symbol: Prickle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020123.1 | - | 1 | 749 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | thyrotropin releasing hormone | Trh |
Pro-TRH, Trf |
-235.8kb | Sense | Chr. 6 - 92242061 - 92244650 on NCBI Build GRCm38 |
Downstream |
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 9 |
Adamts9 |
1810011L16Rik, 8430403M15Rik, AW743315, E030027K14Rik |
291.3kb | Sense | Chr. 6 - 92772699 - 92944047 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
