Assay Details
| Gene Symbol: 9130017K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020265.1 | - | 1 | 1853 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin specific peptidase 43 | Usp43 | -25.0kb | Anti-sense | Chr. 11 - 67854523 - 67922153 on NCBI Build GRCm38 | |
| Downstream | syntaxin 8 | Stx8 |
0610007H08Rik, 1110002H11Rik, 4930571E13Rik, CARB |
17.1kb | Sense | Chr. 11 - 67966411 - 68207148 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9130017K11Rik
Genomic Map