Assay Details
Gene Symbol: 9230112J17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040463.1 | 5 | 2075 | 92 | ||
NR_040464.1 | 4 | 1960 | 92 | |||
GenBank mRNA | AK020339.1 | - | 1 | 55 | 92 | |
AK038653.1 | - | 4 | 1964 | 92 | ||
BB174628.2 | - | 1 | 306 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
thrombospondin, type I, domain containing 4 |
Thsd4 |
ADAMTSL-6, ADAMTSL6, AI585901, Adamtsl4, B230114P05Rik |
-1.5kb | Sense | Chr. 9 - 59966931 - 60522171 on NCBI Build GRCm38 |
Downstream | leucine rich repeat containing 49 | Lrrc49 |
AI430817, AW050057, D430025H09Rik, PGs4, p79 |
62.8kb | Sense | Chr. 9 - 60587229 - 60688134 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2422685 | AK038653.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2422685 | BB174628.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744461 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2860 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304697 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889921 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254533 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807471 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695694 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823371 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195573 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340976 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93416 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2422685 | AK038653.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2422685 | BB174628.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06809468 | - | - |
Whole Mouse Genome G4122A | A_52_P298053 | AK038653.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P298053 | BB174628.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P298053 | AK020339.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124641 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06809468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07589292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559495 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07473566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559585 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559603 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08151826 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07896470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559526 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07727289 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559527 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559428 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07970842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559588 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07588761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07970846 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559553 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07970847 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559500 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08073285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559592 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08167442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07896471 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08073290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559521 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08165179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07588765 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08181427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08167440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559488 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559597 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07730537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08049888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07652607 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559591 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26559567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08029480 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07651582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08073282 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1432867_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454392_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1454392_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1454392_at | - | - |
Mouse Genome 430 2.0 Array | 1432867_at | - | - |
Mouse Genome 430 2.0 Array | 1432867_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900020404.mm.1 | NR_040463.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900020404.mm.1 | NR_040463.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900020404.mm.1 | NR_040464.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900020404.mm.1 | NR_040464.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900020404.mm.1 | TC0900002434.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900020405.mm.1 | NR_040463.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900020405.mm.1 | NR_040463.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900020405.mm.1 | NR_040464.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900020405.mm.1 | NR_040464.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900020405.mm.1 | TC0900002434.mm.1 | MTA TC |
Mouse Expression Set 430 | 1432867_at | - | - |
Mouse Expression Set 430 | 1454392_PM_at | - | - |
Mouse Expression Set 430 | 1454392_at | - | - |
Mouse Expression Set 430 | 1432867_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454392_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432867_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454392_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432867_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 346498 | - | - |
