Assay Details
| Gene Symbol: 9330109K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020364.1 | - | 1 | 352 | 120 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | a disintegrin and metallopeptidase domain 29 | Adam29 | -222.7kb | Anti-sense | Chr. 8 - 55870912 - 55906964 on NCBI Build GRCm38 | |
| Downstream | hydroxyprostaglandin dehydrogenase 15 (NAD) | Hpgd |
15-PGDH, AV026552 |
164.5kb | Sense | Chr. 8 - 56294552 - 56321046 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9330109K16Rik
Genomic Map