Assay Details
Gene Symbol: Pxylp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020522.1 | - | 345 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA C430002N11 gene | C430002N11Rik | -48.4kb | Anti-sense | Chr. 9 - 96765562 - 96774397 on NCBI Build GRCm38 | |
Downstream | splA/ryanodine receptor domain and SOCS box containing 4 | Spsb4 |
D030068E18Rik, SSB-4, Ssb4 |
120.0kb | Sense | Chr. 9 - 96943482 - 97018355 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
