Assay Details
Gene Symbol: Zfp719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020523.1 | - | 1 | 554 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 658 | Zfp658 | -3.6kb | Sense | Chr. 7 - 43562370 - 43576142 on NCBI Build GRCm38 | |
Downstream | zinc finger protein 819 | Zfp819 |
4930427I11Rik, 4933405K07Rik |
26.7kb | Sense | Chr. 7 - 43607169 - 43618279 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
