Assay Details
Gene Symbol: 9530001N24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020533.1 | - | 1 | 253 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | expressed sequence AV051173 | AV051173 | -11.1kb | Anti-sense | Chr. 4 - 116684962 - 116686022 on NCBI Build GRCm38 | |
Downstream |
methylmalonic aciduria cblC type, with homocystinuria |
Mmachc | 1810037K07Rik | 4.9kb | Anti-sense | Chr. 4 - 116702434 - 116708385 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
