Assay Details
| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020687.1 | - | 1 | 553 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sirtuin 2 | Sirt2 |
5730427M03Rik, SIR2L2, Sir2l |
-307.0 | Sense | Chr. 7 - 28766752 - 28788665 on NCBI Build GRCm38 |
| Downstream | heterogeneous nuclear ribonucleoprotein L | Hnrnpl |
C79783, D830027H13Rik, Hnrpl |
19.0kb | Sense | Chr. 7 - 28808546 - 28822268 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rinl
Genomic Map