Assay Details
Gene Symbol: 9930120I10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020689.1 | - | 1 | 651 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glycoprotein 9 (platelet) | Gp9 |
Cd42, GPIX |
-25.7kb | Anti-sense | Chr. 6 - 87776122 - 87779786 on NCBI Build GRCm38 |
Downstream | ISY1 splicing factor homolog (S. cerevisiae) | Isy1 |
5830446M03Rik, AI181014, AU020769 |
12.1kb | Sense | Chr. 6 - 87818447 - 87838759 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
