Assay Details
| Gene Symbol: Ctso | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020735.1 | - | 1 | 649 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A830029E22 gene | A830029E22Rik | -127.5kb | Sense | Chr. 3 - 81801967 - 81805162 on NCBI Build GRCm38 | |
| Downstream | tryptophan 2,3-dioxygenase | Tdo2 |
AA407491, TDO, TO, chky |
25.1kb | Anti-sense | Chr. 3 - 81958412 - 81975728 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctso
Genomic Map