Assay Details
| Gene Symbol: Gm12108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032705.1 | - | 1 - 2 | 87 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuron specific gene family member 2 | Nsg2 | -36.1kb | Anti-sense | Chr. 11 - 32000386 - 32059211 on NCBI Build GRCm38 | |
| Downstream | interleukin 9 receptor | Il9r | 86.3kb | Sense | Chr. 11 - 32187544 - 32205340 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12108
Genomic Map