Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1981276 |
AK131772.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
191451 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
894469 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344481 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
314250 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
791419 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
266511 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
395411 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
290622 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897952 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
488808 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
327717 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889394 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
873702 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
287313 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
161373 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654358 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
78269 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
202748 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
509029 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
357247 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
498566 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
413518 |
AK016451.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
20186 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
250662 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
669461 |
AK010286.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
669461 |
NM_011975.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
126866 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
972364 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
169367 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
766929 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
21380 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
177896 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
178975 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
73899 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
128577 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
175723 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
5836 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1981276 |
AK131772.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377704 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377688 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377733 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377669 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377745 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377620 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377643 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377729 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377593 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08377613 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08377643 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08377688 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08377600 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240135 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240124 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240115 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240117 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240116 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240121 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240134 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240129 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240123 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240139 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240132 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240137 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240131 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240127 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30240119 |
- |
- |