Assay Details
Gene Symbol: A930024N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK044580.1 | - | 1 - 2 | 364 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 10510 | Gm10510 | ENSMUSG00000073453 | -20.5kb | Anti-sense | Chr. 17 - 15350813 - 15354461 on NCBI Build GRCm38 |
Downstream |
family with sequence similarity 120, member B |
Fam120b |
4932442K08Rik, AW049283, AW413338, CCPG, Pgcc1, mKIAA1838 |
19.8kb | Sense | Chr. 17 - 15396157 - 15433581 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
