Assay Details
| Gene Symbol: Gm34245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029797.1 | - | 1 | 2044 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
neurotrophic tyrosine kinase, receptor, type 2 |
Ntrk2 |
GP145-TrkB/GP95-TrkB, Tkrb, trk-B, trkB |
Anti-sense | Chr. 13 - 58806569 - 59133970 on NCBI Build GRCm38 | |
| Downstream | ATP/GTP binding protein 1 | Agtpbp1 |
1700020N17Rik, 2310001G17Rik, 2900054O13Rik, 4930445M19Rik, 5730402G09Rik, CCP1, Nna1, nmf243, pcd |
371.2kb | Sense | Chr. 13 - 59449534 - 59557364 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm34245
Genomic Map