Assay Details
Gene Symbol: Gm26512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029834.1 | - | 3 - 4 | 371 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
deiodinase, iodothyronine, type II |
Dio2 |
5DII, AI324267, DIOII |
-6.7kb | Anti-sense | Chr. 12 - 90724552 - 90738438 on NCBI Build GRCm38 |
Downstream | centrosomal protein 128 | Cep128 |
4930534B04Rik, 5430424K18Rik, AI449708, AI480494, E230024F20 |
247.8kb | Anti-sense | Chr. 12 - 90998490 - 91384416 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
