Assay Details
Gene Symbol: Gm27198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK133101.1 | - | 2 - 3 | 168 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
ATP-binding cassette, sub-family C (CFTR/MRP), member 4 |
Abcc4 |
D630049P08Rik, MOATB, MRP4 |
-20.2kb | Sense | Chr. 14 - 118482692 - 118707620 on NCBI Build GRCm38 |
Downstream | claudin 10 | Cldn10 |
6720456I16Rik, Cldn10a, Cldn10b, D14Ertd728e |
58.5kb | Anti-sense | Chr. 14 - 118787871 - 118874525 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
