Assay Details
Gene Symbol: Gm35853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138921.1 | - | 780 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | keratin 80 | Krt80 |
1200016G03Rik, 2310041I20Rik, Kb20 |
-33.3kb | Anti-sense | Chr. 15 - 101348315 - 101370125 on NCBI Build GRCm38 |
Downstream | keratin 7 | Krt7 |
D15Wsu77e, K7, Krt2-7 |
8.2kb | Sense | Chr. 15 - 101412403 - 101427806 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
