Assay Details
| Gene Symbol: C230037L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036478.1 | - | 1 | 447 | 75 | |
| AK082325.1 | - | 1 | 427 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase 8 interacting protein 2 | Mapk8ip2 |
3230402N03Rik, AI847694, AW049958, IB2, Jip2, R75148 |
-13.8kb | Sense | Chr. 15 - 89453911 - 89462447 on NCBI Build GRCm38 |
| Downstream | SH3/ankyrin domain gene 3 | Shank3 |
AI841104, SHANK3c-3, SHANK3c-4, Shank3b |
22.9kb | Sense | Chr. 15 - 89499707 - 89560261 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C230037L18Rik
Genomic Map