Assay Details
| Gene Symbol: Ntf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037915.1 | - | 1 | 554 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
potassium voltage-gated channel, shaker-related subfamily, member 7 |
Kcna7 | Kv1.7 | -2.5kb | Sense | Chr. 7 - 45405960 - 45411382 on NCBI Build GRCm38 |
| Downstream | luteinizing hormone beta | Lhb |
LH, LH-B, LSH-B, LSH-beta |
3.6kb | Sense | Chr. 7 - 45418166 - 45421855 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ntf5
Genomic Map