Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600025171.mm.1 |
TC0600003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025171.mm.1 |
NR_045009.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025171.mm.1 |
NR_045009.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025174.mm.1 |
TC0600003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025174.mm.1 |
NR_045009.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025174.mm.1 |
NR_045009.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025176.mm.1 |
TC0600003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025176.mm.1 |
NR_045009.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025176.mm.1 |
NR_045009.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025173.mm.1 |
TC0600003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025173.mm.1 |
NR_045009.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025173.mm.1 |
NR_045009.2 |
Refseq |