Assay Details
| Gene Symbol: Gm13982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040062.1 | - | 1 - 2 | 100 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RAS guanyl releasing protein 1 | Rasgrp1 | Rasgrp | -72.2kb | Sense | Chr. 2 - 117279993 - 117342877 on NCBI Build GRCm38 | 
| Downstream | predicted gene 13985 | Gm13985 | OTTMUSG00000015235 | 387.0kb | Sense | Chr. 2 - 117804649 - 117812802 on NCBI Build GRCm38 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length greater than or equal to 101  3' Most  Mus musculus  1700  Probe spans exons | 
MicroArray Probes:
 Back To Top
 Back To Top
 Gm13982
Gm13982 Genomic Map
Genomic Map Back To Top
 Back To Top