Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800015983.mm.1 |
NR_040376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015983.mm.1 |
NR_040376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015983.mm.1 |
TC0800002054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800015985.mm.1 |
NR_040376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015985.mm.1 |
NR_040376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015985.mm.1 |
TC0800002054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800015986.mm.1 |
NR_040376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015986.mm.1 |
NR_040376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015986.mm.1 |
TC0800002054.mm.1 |
MTA TC |