Assay Details
| Gene Symbol: Gm33609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040435.1 | - | 1 - 2 | 159 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | progestin and adipoQ receptor family member III | Paqr3 |
6330415A20Rik, AY424292, RKTG |
-31.3kb | Anti-sense | Chr. 5 - 97082329 - 97111748 on NCBI Build GRCm38 |
| Downstream |
N(alpha)-acetyltransferase 11, NatA catalytic subunit |
Naa11 |
Ard1b, Ard2, C80008 |
234.5kb | Anti-sense | Chr. 5 - 97382209 - 97392330 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm33609
Genomic Map