Assay Details
Gene Symbol: Gm19782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045071.1 | 2 - 3 | 213 | 81 | ||
GenBank mRNA | AK139312.1 | - | 2 - 3 | 215 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930504C09 gene | 4930504C09Rik | -697.0kb | Sense | Chr. 15 - 69270541 - 69274718 on NCBI Build GRCm38 | |
Downstream |
family with sequence similarity 135, member B |
Fam135b |
1700010C24Rik, A830008O07Rik |
1.4mb | Anti-sense | Chr. 15 - 71438951 - 71728436 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
