Assay Details
Gene Symbol: Gm19782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045071.1 | 5 - 6 | 482 | 91 | ||
GenBank mRNA | AK139312.1 | - | 5 - 6 | 484 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930504C09 gene | 4930504C09Rik | -798.4kb | Sense | Chr. 15 - 69270541 - 69274718 on NCBI Build GRCm38 | |
Downstream |
family with sequence similarity 135, member B |
Fam135b |
1700010C24Rik, A830008O07Rik |
1.4mb | Anti-sense | Chr. 15 - 71438951 - 71728436 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261046 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787887 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509091 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614223 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652457 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 792461 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810059 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966931 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423852 | AF216873.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423852 | AK028586.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423852 | AK029014.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423852 | AK029262.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423852 | AK053877.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423852 | AK093831.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423852 | BC040790.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423852 | BC051432.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423852 | NM_019811.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525051 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446647 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472397 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525088 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713659 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602337 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371038 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376881 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306579 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591513 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410300 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645742 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716665 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969875 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902733 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162009 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51463 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509710 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832044 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323750 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282676 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734544 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48527 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884107 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662429 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59396 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413027 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88624 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293517 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688743 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3589 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825349 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811122 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827185 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714201 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517223 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303744 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38400 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45980 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159502 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16960 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19207 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60629 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4177 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134410 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100519 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407160 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03063335 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407240 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407302 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407101 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407260 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407118 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03063364 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407201 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407066 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407140 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08407044 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03063364 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08407044 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08407160 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08407302 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08407101 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12078479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311114 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12078483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311123 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311573 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311094 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311571 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12291387 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311109 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12028727 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12291394 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12028735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30311578 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1500003566.mm.1 | NR_045071.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500003566.mm.1 | NR_045071.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500003566.mm.1 | TC1500000506.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500003565.mm.1 | NR_045071.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500003565.mm.1 | NR_045071.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500003565.mm.1 | TC1500000506.mm.1 | MTA TC |
