Assay Details
Gene Symbol: Gm16239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139491.1 | - | 2 - 3 | 248 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 4 | Galnt4 | AV011803 | -66.8kb | Anti-sense | Chr. 10 - 99108135 - 99113247 on NCBI Build GRCm38 |
Downstream | dual specificity phosphatase 6 | Dusp6 |
1300019I03Rik, MKP-3, MKP3, PYST1 |
80.9kb | Anti-sense | Chr. 10 - 99263231 - 99267489 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
