Assay Details
| Gene Symbol: Gm38973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146287.1 | - | 1 - 2 | 68 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thymoma viral proto-oncogene 2 | Akt2 |
2410016A19Rik, AW554154, PKB, PKBbeta |
-10.0kb | Anti-sense | Chr. 7 - 27591557 - 27639453 on NCBI Build GRCm38 |
| Downstream | tetratricopeptide repeat domain 9B | Ttc9b | 2900074C18Rik | 2.9kb | Anti-sense | Chr. 7 - 27653924 - 27656207 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm38973
Genomic Map