Assay Details
Gene Symbol: LOC102633321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK165035.1 | - | 1 - 2 | 674 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 39329 |
Gm39329 | Anti-sense | Chr. 9 - 46320044 - 46331635 on NCBI Build GRCm38 | ||
Downstream | RIKEN cDNA 2900052N01 gene | 2900052N01Rik | AU067665 | 584.2kb | Anti-sense | Chr. 9 - 46913603 - 46927366 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
