Assay Details
| Gene Symbol: Stamos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036096.1 | - | 1 | 325 | 83 | |
| AK050585.1 | - | 1 | 354 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | 3-hydroxyacyl-CoA dehydratase 1 | Hacd1 | Ptpla | -17.4kb | Sense | Chr. 2 - 14026366 - 14056035 on NCBI Build GRCm38 |
| Downstream | transmembrane protein 236 | Tmem236 |
2010003H20Rik, Fam23a, Gm613 |
100.7kb | Anti-sense | Chr. 2 - 14174524 - 14221993 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stamos
Genomic Map