Assay Details
| Gene Symbol: Gm16253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052161.1 | - | 1 - 2 | 217 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15441 | Gm15441 |
ENSMUSG00000074398, OTTMUSG00000021867 |
-10.2kb | Anti-sense | Chr. 3 - 96555768 - 96566801 on NCBI Build GRCm38 |
| Downstream | ankyrin repeat domain 34A | Ankrd34a |
AW060394, Ankrd34 |
14.2kb | Sense | Chr. 3 - 96594826 - 96600126 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm16253
Genomic Map