Assay Details
| Gene Symbol: Chst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053064.1 | - | 1 | 3044 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium channel tetramerisation domain containing 15 | Kctd15 | BC031749 | -38.8kb | Sense | Chr. 7 - 34639015 - 34655507 on NCBI Build GRCm38 |
| Downstream | peptidase D | Pepd |
Pep-4, Pep4, dal |
215.0kb | Anti-sense | Chr. 7 - 34912407 - 35044708 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chst8
Genomic Map